Assay Details
| Gene Symbol: hsd17b12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200881.1 | NP_957175.1 | 2 - 3 | 346 | 78 | |
| GenBank mRNA | AY551082.1 | - | 2 - 3 | 339 | 78 | |
| BC048053.1 | - | 2 - 3 | 288 | 78 | ||
| BC063943.1 | - | 2 - 3 | 346 | 78 | ||
| BC164345.1 | - | 2 - 3 | 263 | 78 | ||
More Information
Set Membership: |
|

hsd17b12a
Genomic Map