Assay Details
| Gene Symbol: rabgef1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200941.1 | NP_957235.1 | 8 - 9 | 1170 | 74 | |
| GenBank mRNA | BC045469.1 | - | 8 - 9 | 1170 | 74 | |
More Information
Set Membership: |
|

rabgef1l
Genomic Map