Assay Details
| Gene Symbol: col1a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201478.1 | NP_958886.1 | 43 - 44 | 3257 | 149 | |
| XM_005155976.4 | XP_005156033.1 | 43 - 44 | 3300 | 149 | ||
| XM_005155980.4 | XP_005156037.1 | 41 - 42 | 3141 | 149 | ||
| GenBank mRNA | AJ318214.1 | - | 2 - 3 | 134 | 149 | |
| BC058045.1 | - | 43 - 44 | 3257 | 149 | ||
| BC066384.1 | - | 43 - 44 | 3246 | 149 | ||
More Information
Set Membership: |
|

col1a1b
Genomic Map