Assay Details
| Gene Symbol: col1a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201478.1 | NP_958886.1 | 6 - 7 | 595 | 150 | |
| XM_005155976.4 | XP_005156033.1 | 6 - 7 | 638 | 150 | ||
| XM_005155980.4 | XP_005156037.1 | 6 - 7 | 641 | 150 | ||
| GenBank mRNA | BC058045.1 | - | 6 - 7 | 595 | 150 | |
More Information
Set Membership: |
|

col1a1b
Genomic Map