Assay Details
| Gene Symbol: pnp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205655.2 | NP_991218.2 | 5 - 6 | 966 | 61 | |
| GenBank mRNA | BC065621.1 | - | 5 - 6 | 922 | 61 | |
| CK028065.1 | - | 5 - 6 | 797 | 61 | ||
More Information
Set Membership: |
|

pnp6
Genomic Map