Assay Details
| Gene Symbol: ctsla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212584.1 | NP_997749.1 | 1 - 2 | 137 | 157 | |
| XM_005165268.2 | XP_005165325.1 | 2 - 3 | 275 | 157 | ||
| XM_021476007.1 | XP_021331682.1 | 2 - 3 | 484 | 157 | ||
| GenBank mRNA | BC066490.1 | - | 1 - 2 | 137 | 157 | |
More Information
Set Membership: |
|

ctsla
Genomic Map