Assay Details
| Gene Symbol: ccnt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212586.1 | NP_997751.1 | 2 - 3 | 270 | 91 | |
| GenBank mRNA | BC048886.1 | - | 2 - 3 | 270 | 91 | |
| BC164276.1 | - | 2 - 3 | 270 | 91 | ||
More Information
Set Membership: |
|

ccnt2b
Genomic Map