Assay Details
| Gene Symbol: hnrnph1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212589.2 | NP_997754.2 | 10 - 11 | 1251 | 63 | |
| XM_021468753.1 | XP_021324428.1 | 9 - 10 | 1147 | 63 | ||
| GenBank mRNA | BC044161.1 | - | 10 - 11 | 1244 | 63 | |
| BC067720.1 | - | 8 - 9 | 972 | 63 | ||
| BC164507.1 | - | 9 - 10 | 1176 | 63 | ||
| CT671502.2 | - | 5 - 6 | 525 | 63 | ||
More Information
Set Membership: |
|

hnrnph1l
Genomic Map