Assay Details
| Gene Symbol: hnrnph1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212589.2 | NP_997754.2 | 3 - 4 | 340 | 76 | |
| XM_021468753.1 | XP_021324428.1 | 3 - 4 | 375 | 76 | ||
| GenBank mRNA | BC044161.1 | - | 3 - 4 | 333 | 76 | |
| BC067720.1 | - | 1 - 2 | 60 | 76 | ||
| BC164507.1 | - | 2 - 3 | 265 | 76 | ||
| EB830940.1 | - | 3 - 4 | 344 | 76 | ||
| EH437757.1 | - | 3 - 4 | 362 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC067720.1, XM_021468753.1 transcripts.
Set Membership: |
|

hnrnph1l
Genomic Map