Assay Details
| Gene Symbol: ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212688.1 | NP_997853.1 | 6 - 7 | 488 | 70 | |
| GenBank mRNA | BC067615.1 | - | 6 - 7 | 488 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ctsh| Gene Symbol: ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212688.1 | NP_997853.1 | 6 - 7 | 488 | 70 | |
| GenBank mRNA | BC067615.1 | - | 6 - 7 | 488 | 70 | |
Set Membership: |
|