Assay Details
| Gene Symbol: ranbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212766.2 | NP_997931.2 | 4 - 5 | 599 | 82 | |
| GenBank mRNA | AF364085.1 | - | 4 - 5 | 532 | 82 | |
| BC067558.1 | - | 4 - 5 | 497 | 82 | ||
| EH549160.1 | - | 4 - 5 | 615 | 82 | ||
| EH562192.1 | - | 2 - 3 | 684 | 82 | ||
More Information
Set Membership: |
|

ranbp1
Genomic Map