Assay Details
| Gene Symbol: traf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212817.1 | NP_997982.1 | 2 - 3 | 311 | 89 | |
| GenBank mRNA | AJ556549.1 | - | 2 - 3 | 408 | 89 | |
More Information
Set Membership: |
|

traf4b
Genomic Map