Assay Details
| Gene Symbol: traf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212817.1 | NP_997982.1 | 4 - 5 | 595 | 76 | |
| GenBank mRNA | AJ556549.1 | - | 4 - 5 | 692 | 76 | |
More Information
Set Membership: |
|

traf4b
Genomic Map