Assay Details
| Gene Symbol: traf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212817.1 | NP_997982.1 | 6 - 7 | 901 | 74 | |
| GenBank mRNA | AJ556549.1 | - | 6 - 7 | 998 | 74 | |
| BC078361.1 | - | 6 - 7 | 810 | 74 | ||
| BC165750.1 | - | 6 - 7 | 809 | 74 | ||
More Information
Set Membership: |
|

traf4b
Genomic Map