Assay Details
| Gene Symbol: arih1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212923.1 | NP_998088.1 | 11 - 12 | 1201 | 69 | |
| GenBank mRNA | BC067684.1 | - | 11 - 12 | 1201 | 69 | |
More Information
Set Membership: |
|

arih1l
Genomic Map