Assay Details
| Gene Symbol: pik3r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291329.1 | NP_001278258.1 | 3 - 4 | 314 | 67 | |
| NM_213038.2 | NP_998203.2 | 2 - 3 | 477 | 67 | ||
| GenBank mRNA | BC059572.1 | - | 2 - 3 | 458 | 67 | |
| CB359701.1 | - | 2 - 3 | 481 | 67 | ||
| EH456435.1 | - | 3 - 4 | 349 | 67 | ||
More Information
Set Membership: |
|

pik3r3a
Genomic Map