Assay Details
| Gene Symbol: ptp4a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213181.1 | NP_998346.1 | 3 - 4 | 809 | 106 | |
| XM_005170023.1 | XP_005170080.1 | 4 - 5 | 819 | 106 | ||
| XM_005170024.4 | XP_005170081.1 | 4 - 5 | 848 | 106 | ||
| GenBank mRNA | BC067186.1 | - | 3 - 4 | 809 | 106 | |
More Information
Set Membership: |
|

ptp4a3a
Genomic Map