Assay Details
| Gene Symbol: m6pr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213205.1 | NP_998370.1 | 3 - 4 | 562 | 116 | |
| GenBank mRNA | BC065858.1 | - | 3 - 4 | 562 | 116 | |
| DQ089037.1 | - | 3 - 4 | 562 | 116 | ||
More Information
Set Membership: |
|

m6pr
Genomic Map