Assay Details
| Gene Symbol: aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213225.1 | NP_998390.1 | 4 - 5 | 385 | 62 | |
| GenBank mRNA | BC067671.1 | - | 4 - 5 | 385 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aaas| Gene Symbol: aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213225.1 | NP_998390.1 | 4 - 5 | 385 | 62 | |
| GenBank mRNA | BC067671.1 | - | 4 - 5 | 385 | 62 | |
Set Membership: |
|