Assay Details
| Gene Symbol: sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213278.1 | NP_998443.1 | 4 - 5 | 1357 | 74 | |
| GenBank mRNA | AY648751.1 | - | 4 - 5 | 1361 | 74 | |
| BC068428.1 | - | 4 - 5 | 1357 | 74 | ||
More Information
Set Membership: |
|

sfpq
Genomic Map