Assay Details
| Gene Symbol: scamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213378.2 | NP_998543.1 | 6 - 7 | 726 | 104 | |
| GenBank mRNA | BC046051.1 | - | 6 - 7 | 703 | 104 | |
| BC066575.1 | - | 6 - 7 | 687 | 103 | ||
| EH539419.1 | - | 6 - 7 | 654 | 104 | ||
More Information
Set Membership: |
|

scamp2
Genomic Map