Assay Details
| Gene Symbol: stk38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213456.1 | NP_998621.1 | 3 - 4 | 242 | 112 | |
| GenBank mRNA | BC044428.1 | - | 3 - 4 | 242 | 112 | |
| BC165742.1 | - | 2 - 3 | 209 | 112 | ||
More Information
Set Membership: |
|

stk38a
Genomic Map