Assay Details
| Gene Symbol: mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213473.1 | NP_998638.1 | 4 - 5 | 865 | 60 | |
| XM_005167152.4 | XP_005167209.1 | 4 - 5 | 966 | 60 | ||
| GenBank mRNA | BC044411.1 | - | 4 - 5 | 865 | 60 | |
| BC164564.1 | - | 2 - 3 | 719 | 60 | ||
| DQ360073.1 | - | 2 - 3 | 685 | 60 | ||
More Information
Set Membership: |
|

mapk4
Genomic Map