Assay Details
| Gene Symbol: gnb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213481.1 | NP_998646.1 | 4 - 5 | 531 | 80 | |
| XM_009302712.2 | XP_009300987.1 | 4 - 5 | 80 | |||
| XM_009302713.2 | XP_009300988.1 | 4 - 5 | 80 | |||
| XM_009302714.2 | XP_009300989.1 | 4 - 5 | 80 | |||
| GenBank mRNA | BC044482.1 | - | 4 - 5 | 531 | 80 | |
| BC152684.1 | - | 4 - 5 | 337 | 80 | ||
| BC164143.1 | - | 3 - 4 | 235 | 80 | ||
More Information
Set Membership: |
|

gnb1b
Genomic Map