Assay Details
| Gene Symbol: gnb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213481.1 | NP_998646.1 | 10 - 11 | 1389 | 79 | |
| XM_009302712.2 | XP_009300987.1 | 10 - 11 | 1213 | 79 | ||
| GenBank mRNA | BC044482.1 | - | 10 - 11 | 1389 | 79 | |
More Information
Set Membership: |
|

gnb1b
Genomic Map