Assay Details
| Gene Symbol: sap30l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214703.1 | NP_999868.1 | 3 - 4 | 479 | 72 | |
| GenBank mRNA | BC066447.1 | - | 3 - 4 | 479 | 72 | |
More Information
Set Membership: |
|

sap30l
Genomic Map