Assay Details
| Gene Symbol: sdr16c5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214771.2 | NP_999936.1 | 2 - 3 | 464 | 66 | |
| GenBank mRNA | BC054138.1 | - | 2 - 3 | 466 | 66 | |
More Information
Set Membership: |
|

sdr16c5b
Genomic Map