Assay Details
| Gene Symbol: asic4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214786.1 | NP_999951.1 | 6 - 7 | 1538 | 104 | |
| GenBank mRNA | AJ609620.1 | - | 6 - 7 | 1538 | 104 | |
| BC095568.1 | - | 6 - 7 | 1545 | 104 | ||
More Information
Set Membership: |
|

asic4b
Genomic Map