Assay Details
| Gene Symbol: fabp7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214807.1 | NP_999972.1 | 2 - 3 | 306 | 90 | |
| GenBank mRNA | AY380814.1 | - | 2 - 3 | 306 | 90 | |
| BC162199.1 | - | 2 - 3 | 293 | 90 | ||
| BC162202.1 | - | 2 - 3 | 293 | 90 | ||
More Information
Set Membership: |
|

fabp7b
Genomic Map