Assay Details
| Gene Symbol: tspan18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002439.1 | NP_001002439.1 | 6 - 7 | 677 | 112 | |
| XM_005159172.4 | XP_005159229.1 | 6 - 7 | 956 | 112 | ||
| XM_005159173.4 | XP_005159230.1 | 6 - 7 | 566 | 112 | ||
| XM_017351914.1 | XP_017207403.1 | 5 - 6 | 791 | 112 | ||
| GenBank mRNA | BC076124.1 | - | 6 - 7 | 677 | 112 | |
More Information
Set Membership: |
|

tspan18b
Genomic Map