Assay Details
| Gene Symbol: ldhbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002474.1 | NP_001002474.1 | 4 - 5 | 583 | 87 | |
| XM_005170658.4 | XP_005170715.1 | 4 - 5 | 691 | 87 | ||
| XM_005170659.2 | XP_005170716.1 | 4 - 5 | 87 | |||
| XM_021470477.1 | XP_021326152.1 | 4 - 5 | 87 | |||
| GenBank mRNA | BC076340.1 | - | 4 - 5 | 583 | 87 | |
| BC165286.1 | - | 3 - 4 | 457 | 87 | ||
More Information
Set Membership: |
|

ldhbb
Genomic Map