Assay Details
| Gene Symbol: sdhda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002489.2 | NP_001002489.1 | 1 - 2 | 176 | 83 | |
| GenBank mRNA | BC076319.1 | - | 1 - 2 | 167 | 83 | |
| BC163980.1 | - | 1 - 2 | 84 | 83 | ||
More Information
Set Membership: |
|

sdhda
Genomic Map