Assay Details
| Gene Symbol: taf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002548.1 | NP_001002548.1 | 4 - 5 | 476 | 85 | |
| XM_005161895.3 | XP_005161952.1 | 4 - 5 | 572 | 85 | ||
| XM_021469778.1 | XP_021325453.1 | 4 - 5 | 548 | 85 | ||
| GenBank mRNA | BC076258.1 | - | 4 - 5 | 476 | 85 | |
| BC154290.1 | - | 4 - 5 | 466 | 85 | ||
More Information
Set Membership: |
|

taf11
Genomic Map