Assay Details
| Gene Symbol: ndufab1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003418.1 | NP_001003418.1 | 1 - 2 | 188 | 72 | |
| GenBank mRNA | BC076098.1 | - | 1 - 2 | 188 | 72 | |
| BC152172.1 | - | 1 - 2 | 188 | 72 | ||
More Information
Set Membership: |
|

ndufab1a
Genomic Map