Assay Details
| Gene Symbol: nutf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003598.2 | NP_001003598.1 | 2 - 3 | 320 | 67 | |
| GenBank mRNA | BC078197.1 | - | 2 - 3 | 229 | 67 | |
| EH475711.1 | - | 3 - 4 | 506 | 67 | ||
| EH518961.1 | - | 3 - 4 | 572 | 67 | ||
| EH538050.1 | - | 2 - 3 | 358 | 67 | ||
More Information
Set Membership: |
|

nutf2l
Genomic Map