Assay Details
| Gene Symbol: riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003614.2 | NP_001003614.1 | 3 - 4 | 536 | 70 | |
| GenBank mRNA | BC077112.1 | - | 3 - 4 | 540 | 70 | |
More Information
Set Membership: |
|

riok3
Genomic Map