Assay Details
| Gene Symbol: alg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003784.2 | NP_001003784.2 | 4 - 5 | 446 | 76 | |
| GenBank mRNA | BC079496.1 | - | 4 - 5 | 450 | 76 | |
| EH513372.1 | - | 4 - 5 | 472 | 76 | ||
More Information
Set Membership: |
|

alg6
Genomic Map