Assay Details
| Gene Symbol: bud31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003860.1 | NP_001003860.1 | 2 - 3 | 304 | 103 | |
| GenBank mRNA | AY648727.1 | - | 2 - 3 | 304 | 103 | |
More Information
Set Membership: |
|

bud31
Genomic Map