Assay Details
| Gene Symbol: syt9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003985.1 | NP_001003985.1 | 1 - 2 | 364 | 66 | |
| GenBank mRNA | BC080256.1 | - | 1 - 2 | 364 | 66 | |
| BC163971.1 | - | 1 - 2 | 183 | 66 | ||
More Information
Set Membership: |
|

syt9b
Genomic Map