Assay Details
| Gene Symbol: pfkma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004575.2 | NP_001004575.1 | 13 - 14 | 1429 | 77 | |
| GenBank mRNA | BC081652.1 | - | 13 - 14 | 1405 | 77 | |
More Information
Set Membership: |
|

pfkma
Genomic Map