Assay Details
| Gene Symbol: pfkma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004575.2 | NP_001004575.1 | 4 - 5 | 520 | 95 | |
| GenBank mRNA | BC081652.1 | - | 4 - 5 | 496 | 95 | |
More Information
Set Membership: |
|

pfkma
Genomic Map