Assay Details
| Gene Symbol: kifap3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004644.1 | NP_001004644.1 | 6 - 7 | 706 | 108 | |
| GenBank mRNA | BC080842.1 | - | 6 - 7 | 706 | 108 | |
More Information
Set Membership: |
|

kifap3a
Genomic Map