Assay Details
| Gene Symbol: gpsm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005936.1 | NP_001005936.1 | 2 - 3 | 374 | 68 | |
| XM_021469788.1 | XP_021325463.1 | 2 - 3 | 400 | 68 | ||
| GenBank mRNA | BC083520.1 | - | 2 - 3 | 374 | 68 | |
| DQ235092.1 | - | 1 - 2 | 221 | 68 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  Mammalian Gene Collection  Recommended Assay  Danio rerio  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 gpsm2l
gpsm2l Genomic Map
Genomic Map Back To Top
 Back To Top