Assay Details
| Gene Symbol: ormdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006087.1 | NP_001006087.1 | 2 - 3 | 481 | 65 | |
| GenBank mRNA | BC083232.1 | - | 2 - 3 | 481 | 65 | |
More Information
Set Membership: |
|

ormdl3
Genomic Map