Assay Details
| Gene Symbol: trpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007066.2 | NP_001007067.1 | 18 - 19 | 2169 | 106 | |
| XM_009297331.3 | XP_009295606.1 | 18 - 19 | 2495 | 106 | ||
| XM_021470156.1 | XP_021325831.1 | 18 - 19 | 2512 | 106 | ||
| GenBank mRNA | AY677197.1 | - | 18 - 19 | 2174 | 106 | |
More Information
Set Membership: |
|

trpa1b
Genomic Map