Assay Details
| Gene Symbol: trpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007066.2 | NP_001007067.1 | 20 - 21 | 2397 | 102 | |
| XM_009297331.3 | XP_009295606.1 | 20 - 21 | 2723 | 102 | ||
| XM_021470156.1 | XP_021325831.1 | 20 - 21 | 2740 | 102 | ||
| GenBank mRNA | AY677197.1 | - | 20 - 21 | 2402 | 102 | |
| EU826642.1 | - | 19 - 20 | 2275 | 102 | ||
More Information
Set Membership: |
|

trpa1b
Genomic Map