Assay Details
| Gene Symbol: trpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007066.2 | NP_001007067.1 | 13 - 14 | 1644 | 71 | |
| XM_009297331.3 | XP_009295606.1 | 13 - 14 | 1970 | 71 | ||
| XM_021470156.1 | XP_021325831.1 | 13 - 14 | 1987 | 71 | ||
| GenBank mRNA | AY677197.1 | - | 13 - 14 | 1649 | 71 | |
| EU826642.1 | - | 12 - 13 | 1522 | 71 | ||
More Information
Set Membership: |
|

trpa1b
Genomic Map