Assay Details
| Gene Symbol: il12ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007108.1 | NP_001007109.1 | 3 - 4 | 431 | 104 | |
| GenBank mRNA | AB183002.1 | - | 3 - 4 | 431 | 104 | |
| BC121764.1 | - | 3 - 4 | 430 | 104 | ||
| BC164577.1 | - | 2 - 3 | 402 | 104 | ||
More Information
Set Membership: |
|

il12ba
Genomic Map