Assay Details
| Gene Symbol: mfsd2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007451.1 | NP_001007452.1 | 5 - 6 | 711 | 100 | |
| XM_005156993.4 | XP_005157050.1 | 6 - 7 | 797 | 100 | ||
| GenBank mRNA | BC085388.1 | - | 5 - 6 | 711 | 100 | |
More Information
Set Membership: |
|

mfsd2aa
Genomic Map