Assay Details
| Gene Symbol: usp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008574.2 | NP_001008574.2 | 2 - 3 | 478 | 65 | |
| XM_005157513.4 | XP_005157570.1 | 4 - 5 | 978 | 65 | ||
| XM_005157514.4 | XP_005157571.1 | 4 - 5 | 1072 | 65 | ||
| XM_005157515.4 | XP_005157572.1 | 4 - 5 | 973 | 65 | ||
| XM_005157516.4 | XP_005157573.1 | 2 - 3 | 546 | 65 | ||
| XM_017350951.2 | XP_017206440.1 | 4 - 5 | 973 | 65 | ||
| XM_021481396.1 | XP_021337071.1 | 4 - 5 | 1076 | 65 | ||
| GenBank mRNA | BC086963.1 | - | 2 - 3 | 319 | 65 | |
More Information
Set Membership: |
|

usp2a
Genomic Map